Stampa…

2-Nitrophenylarsonic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

N0133-1G N0133-25G
TCIAN0133-1GEA 16 EUR
TCIAN0133-1G TCIAN0133-25G
2-Nitrophenylarsonic acid ≥98.0% (by titrimetric analysis)
2-Nitrophenylarsonic acid
Formula: C₆H₆AsNO₅
MDL Number: MFCD00047664
CAS Number: 5410-29-7
EINECS: 226-485-0
UN: 3465
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa07a9719d0b3f436ef62b2f7b1aa739a697d0f&vid=afa07a9719d0b3f436ef62b2f7b1aa739a697d0f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR