Tetraethylene glycol monomethyl ether ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
mPEG4-OH
T1372-25G
T1372-5G
TCIAT1372-25GEA
157
EUR
TCIAT1372-25G
TCIAT1372-5G
Tetraethylene glycol monomethyl ether ≥98.0% (by GC)
Tetraethylene glycol monomethyl ether
Formula:
C₉H₂₀O₅ Peso molecolare: 208,25 g/mol Punto di ebollizione: 125°C |
MDL Number:
MFCD00041756 CAS Number: 23783-42-8 EINECS: 245-883-5 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...