Stampa…

Tetraethylene glycol ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol

T0099-25G T0099-500G
TCIAT0099-25GEA 16 EUR
TCIAT0099-25G TCIAT0099-500G
Tetraethylene glycol ≥95.0% (by GC)
Tetraethylene glycol
Formula: C₈H₁₈O₅
Peso molecolare: 194,23 g/mol
Punto di ebollizione: 328°C
Flash Pt: 194°C
MDL Number: MFCD00002879
CAS Number: 112-60-7
EINECS: 203-989-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47b470a60630e227ee7e0451cba221f282f1d6ec&vid=47b470a60630e227ee7e0451cba221f282f1d6ec&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR