Aminoacetaldehyde dimethyl acetal ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2,2-Dimethoxyethanamine
A0801-25ML
A0801-250ML
TCIAA0801-25MLEA
37
EUR
TCIAA0801-25ML
TCIAA0801-250ML
Aminoacetaldehyde dimethyl acetal ≥98.0% (by GC, titration analysis)
Aminoacetaldehyde dimethyl acetal
Formula:
C₄H₁₁NO₂ Peso molecolare: 105,14 g/mol Punto di ebollizione: 136°C Flash Pt: 53°C |
MDL Number:
MFCD00008135 CAS Number: 22483-09-6 EINECS: 245-026-5 UN: 2924 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...