Diethyl-3-oxoglutarate ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate
A0055-25ML
A0055-500ML
TCIAA0055-25MLEA
37
EUR
TCIAA0055-25ML
TCIAA0055-500ML
Diethyl-3-oxoglutarate ≥95.0% (by GC)
Diethyl-3-oxoglutarate
Formula:
C₉H₁₄O₅ Peso molecolare: 202,21 g/mol Punto di ebollizione: 250°C Flash Pt: 71°C |
MDL Number:
MFCD00009200 CAS Number: 105-50-0 EINECS: 203-302-2 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...