Bis(trimethylsilyl)acetylene ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
B1090-25G
B1090-5G
TCIAB1090-25GEA
246
EUR
TCIAB1090-25G
TCIAB1090-5G
Bis(trimethylsilyl)acetylene ≥97.0% (by GC)
Bis(trimethylsilyl)acetylene
Formula:
C₈H₁₈Si₂ Peso molecolare: 170,4 g/mol Flash Pt: 2°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00008276 CAS Number: 14630-40-1 EINECS: 238-671-9 UN: 1325 ADR: 4.1,II |
Risultati del test di specificazione
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...