Phenylphosphonic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Phenylphosphonic Acid
P0204-25G
P0204-500G
P0204-100G
TCIAP0204-25GEA
21
EUR
TCIAP0204-25G
TCIAP0204-500G
TCIAP0204-100G
Phenylphosphonic acid ≥98.0% (by titrimetric analysis)
Phenylphosphonic acid
Formula:
C₆H₇O₃P Peso molecolare: 158,09 g/mol |
MDL Number:
MFCD00002136 CAS Number: 1571-33-1 EINECS: 216-388-1 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 162.0 to 166.0 deg-C |
Solubility in Water | faint turbidity |
Solubility in Methanol | faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...