Stampa…

Benzylmalonic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

B0422-25G B0422-5G
TCIAB0422-25GEA 158 EUR
TCIAB0422-25G TCIAB0422-5G
Benzylmalonic acid ≥98.0% (by GC, titration analysis)
Benzylmalonic acid
Formula: C₁₀H₁₀O₄
MDL Number: MFCD00004261
CAS Number: 616-75-1
EINECS: 210-491-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b38bbf7c231f0277688bf8a3e65b2dd799ef74e7&vid=b38bbf7c231f0277688bf8a3e65b2dd799ef74e7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR