Stampa…

6-Methoxypicolinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Methoxypyridine-2-carboxylic acid, 6-Methoxy-2-pyridinecarboxylic acid

M2189-1G M2189-5G
TCIAM2189-1GEA 49 EUR
TCIAM2189-1G TCIAM2189-5G
6-Methoxypicolinic acid ≥98.0% (by GC, titration analysis)
6-Methoxypicolinic acid
Formula: C₇H₇NO₃
Peso molecolare: 153,14 g/mol
MDL Number: MFCD06800962
CAS Number: 26893-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9774680787ee5872ed6d9ad3f9e722cd19e46809&vid=9774680787ee5872ed6d9ad3f9e722cd19e46809&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR