Stampa…

3-Methylpicolinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Methylpyridine-2-carboxylic acid

M1141-1G M1141-100MG M1141-5G
TCIAM1141-1GEA 27 EUR
TCIAM1141-1G TCIAM1141-100MG TCIAM1141-5G
3-Methylpicolinic acid ≥98.0% (by GC, titration analysis)
3-Methylpicolinic acid
Formula: C₇H₇NO₂
Peso molecolare: 137,14 g/mol
MDL Number: MFCD00191553
CAS Number: 4021-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d04e27a3388b4e6a2866e98142e64975a390fee5&vid=d04e27a3388b4e6a2866e98142e64975a390fee5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 121.0 deg-C
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR