Stampa…

(±)-Ethyl mandelate ≥95.0% (by GC)

Fornitore: TCI
Attenzione

M0039-25G M0039-500G
TCIAM0039-25GEA 24 EUR
TCIAM0039-25G TCIAM0039-500G
(±)-Ethyl mandelate ≥95.0% (by GC)
(±)-Ethyl mandelate
Formula: C₁₀H₁₂O₃
Punto di ebollizione: 133°C
MDL Number: MFCD00004494
CAS Number: 774-40-3
EINECS: 212-263-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ced04a98b8e186fbf56ae6323ef36b0e1a16c080&vid=ced04a98b8e186fbf56ae6323ef36b0e1a16c080&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Freezing point 23.0 to 27.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR