Stampa…

1,1'-Dimethylferrocene ≥93.0% (by GC)

Fornitore: TCI EUROPE

D1273-25G D1273-5G
TCIAD1273-25GEA 388 EUR
TCIAD1273-25G TCIAD1273-5G
1,1'-Dimethylferrocene ≥93.0% (by GC)
1,1'-Dimethylferrocene
Formula: C₁₂H₁₄Fe
Peso molecolare: 214,09 g/mol
MDL Number: MFCD00001425
CAS Number: 1291-47-0
EINECS: 215-067-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a3103e0e5b4d2721c12eeb92e5d4ba29aeb39e6&vid=7a3103e0e5b4d2721c12eeb92e5d4ba29aeb39e6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to Amber to Dark red powder to crystal
Purity (GC) min. 93.0 %
Melting point 38.0 to 42.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR