6-Fluoronicotinic acid ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Fluoro-5-pyridinecarboxylic acid
F0695-1G
F0695-200MG
F0695-5G
TCIAF0695-1GEA
55
EUR
TCIAF0695-1G
TCIAF0695-200MG
TCIAF0695-5G
6-Fluoronicotinic acid ≥98.0% (by GC)
6-Fluoronicotinic acid
Formula:
C₆H₄FNO₂ Peso molecolare: 141,1 g/mol Punto di fusione: 147°C |
MDL Number:
MFCD01859863 CAS Number: 403-45-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...