6-Fluoropicolinic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Fluoro-6-pyridinecarboxylic acid, 6-Fluoropyridine-2-carboxylic acid
F0837-1G
F0837-5G
TCIAF0837-1GEA
49
EUR
TCIAF0837-1G
TCIAF0837-5G
6-Fluoropicolinic acid ≥98.0% (by titrimetric analysis)
6-Fluoropicolinic acid
Formula:
C₆H₄FNO₂ Peso molecolare: 141,1 g/mol |
MDL Number:
MFCD02181193 CAS Number: 402-69-7 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 136.0 to 140.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...