5-Aminonicotinic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
5-Aminopyridine-3-carboxylic acid, 5-Amino-3-pyridinecarboxylic acid
Formula:
C₆H₆N₂O₂ Peso molecolare: 138.13 g/mol Punto di fusione: 293°C |
MDL Number:
MFCD00129116 CAS Number: 24242-19-1 |
|
Risultati del test di specificazione
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...