Stampa…

6-Bromopicolinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid

B3024-1G B3024-5G
TCIAB3024-1GEA 26 EUR
TCIAB3024-1G TCIAB3024-5G
6-Bromopicolinic acid ≥98.0% (by GC, titration analysis)
6-Bromopicolinic acid
Formula: C₆H₄BrNO₂
Peso molecolare: 202,01 g/mol
MDL Number: MFCD00093197
CAS Number: 21190-87-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aec54766468eb022a45719b26be3bf0cc89bb79c&vid=aec54766468eb022a45719b26be3bf0cc89bb79c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR