Stampa…

5-Bromopicolinic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid

B3188-1G B3188-5G
TCIAB3188-1GEA 18 EUR
TCIAB3188-1G TCIAB3188-5G
5-Bromopicolinic acid ≥97.0% (by GC, titration analysis)
5-Bromopicolinic acid
Formula: C₆H₄BrNO₂
Peso molecolare: 202,01 g/mol
Punto di fusione: 176°C
MDL Number: MFCD00234149
CAS Number: 30766-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38aad293048cc8bef10d103c62f43fcdcffa7ba7&vid=38aad293048cc8bef10d103c62f43fcdcffa7ba7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR