Stampa…

N,N-Dimethylglycine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

D3585-25G D3585-5G
TCIAD3585-25GEA 123 EUR
TCIAD3585-25G TCIAD3585-5G
N,N-Dimethylglycine ≥98.0% (by titrimetric analysis)
N,N-Dimethylglycine
Formula: C₄H₉NO₂
Peso molecolare: 103,12 g/mol
MDL Number: MFCD00004283
CAS Number: 1118-68-9
EINECS: 214-267-8
Merck Index: 14,03244
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=024a2949a1090a30d2991e2396344e08e3b4304e&vid=024a2949a1090a30d2991e2396344e08e3b4304e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 179.0 to 183.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR