Stampa…

4'-Ethoxyacetanilide ≥99.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide

P1669-25G P1669-500G
TCIAP1669-25GEA 28 EUR
TCIAP1669-25G TCIAP1669-500G
4'-Ethoxyacetanilide ≥99.0% (by GC)
4'-Ethoxyacetanilide
Formula: C₁₀H₁₃NO₂
Peso molecolare: 179,22 g/mol
Punto di ebollizione: 132°C
MDL Number: MFCD00009094
CAS Number: 62-44-2
EINECS: 200-533-0
Merck Index: 14,07204
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5fae57d2ab3cb6a6bbd801778e1183d8019b549&vid=e5fae57d2ab3cb6a6bbd801778e1183d8019b549&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 134.0 to 138.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR