(S)-(+)-β-Aminophenethyl alcohol ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol
P1294-25G
P1294-5G
TCIAP1294-25GEA
75
EUR
TCIAP1294-25G
TCIAP1294-5G
(S)-(+)-β-Aminophenethyl alcohol ≥97.0% (by GC, titration analysis)
(S)-(+)-β-Aminophenethyl alcohol
Formula:
C₈H₁₁NO Peso molecolare: 137,18 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00064404 CAS Number: 20989-17-7 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 72.0 to 79.0 deg-C |
Specific rotation [a]20/D | +27.0 to +32.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...