(R)-(-)-β-Aminophenethyl alcohol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
D-Phenylglycinol, (R)-(-)-2-Phenylglycinol
P1201-25G
P1201-5G
TCIAP1201-25GEA
67
EUR
TCIAP1201-25G
TCIAP1201-5G
(R)-(-)-β-Aminophenethyl alcohol ≥98.0% (by GC, titration analysis)
(R)-(-)-β-Aminophenethyl alcohol
Formula:
C₈H₁₁NO Peso molecolare: 137,18 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00008062 CAS Number: 56613-80-0 EINECS: 260-287-5 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 75.0 to 80.0 deg-C |
Specific rotation [a]20/D | -28.0 to -31.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...