Diisopropyl phosphonate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Diisopropyl phosphite, Di-i-propylphosphite, Diisopropyl hydrogen phosphite
P0629-25G
P0629-500G
TCIAP0629-25GEA
26
EUR
TCIAP0629-25G
TCIAP0629-500G
Diisopropyl phosphonate ≥98.0% (by GC)
Diisopropyl phosphonate
Formula:
C₆H₁₅O₃P Peso molecolare: 166,16 g/mol Punto di ebollizione: 79°C Flash Pt: 112°C |
MDL Number:
MFCD00117905 CAS Number: 1809-20-7 EINECS: 217-317-7 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...