3-Chloropicolinic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Chloropyridine-2-carboxylic acid
C2342-1G
C2342-5G
TCIAC2342-1GEA
73
EUR
TCIAC2342-1G
TCIAC2342-5G
3-Chloropicolinic acid ≥98.0% (by GC, titration analysis)
3-Chloropicolinic acid
Formula:
C₆H₄ClNO₂ Peso molecolare: 157,56 g/mol Punto di fusione: 154°C |
MDL Number:
MFCD01646325 CAS Number: 57266-69-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...