Stampa…

2,6-Dichloroisonicotinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,6-Dichloropyridine-4-carboxylic acid

D3345-1G D3345-5G
TCIAD3345-1GEA 25 EUR
TCIAD3345-1G TCIAD3345-5G
2,6-Dichloroisonicotinic acid ≥98.0% (by GC, titration analysis)
2,6-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
Peso molecolare: 192 g/mol
Punto di fusione: 211°C
MDL Number: MFCD00234147
CAS Number: 5398-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f776a39a18a2cad0c41aaf98f50f9310ea61f34f&vid=f776a39a18a2cad0c41aaf98f50f9310ea61f34f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR