6-Chloroisatin ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Coroindoline-2,3-dione
C1871-25G
C1871-5G
TCIAC1871-25GEA
190
EUR
TCIAC1871-25G
TCIAC1871-5G
6-Chloroisatin ≥98.0% (by GC, titration analysis)
6-Chloroisatin
Formula:
C₈H₄ClNO₂ Peso molecolare: 181,58 g/mol Punto di fusione: 262°C |
MDL Number:
MFCD00086347 CAS Number: 6341-92-0 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...