Stampa…

Ethylene glycol monoisopropyl ether ≥99.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Isopropoxyethanol

E0110-25ML E0110-500ML
TCIAE0110-25MLEA 17 EUR
TCIAE0110-25ML TCIAE0110-500ML
Ethylene glycol monoisopropyl ether ≥99.0% (by GC)
Ethylene glycol monoisopropyl ether
Formula: C₅H₁₂O₂
Peso molecolare: 104,15 g/mol
Punto di ebollizione: 141°C
Flash Pt: 46°C
MDL Number: MFCD00002866
CAS Number: 109-59-1
EINECS: 203-685-6
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f81620b64ed7ba98e01f06a2768f0c1e6a17d48&vid=9f81620b64ed7ba98e01f06a2768f0c1e6a17d48&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR