2,7-Dibromofluorene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,7-Dibromo-9H-fluorene
D3556-100G
D3556-25G
D3556-5G
TCIAD3556-100GEA
117
EUR
TCIAD3556-100G
TCIAD3556-25G
TCIAD3556-5G
2,7-Dibromofluorene ≥98.0% (by GC)
2,7-Dibromofluorene
Formula:
C₁₃H₈Br₂ Peso molecolare: 324,01 g/mol |
MDL Number:
MFCD00019048 CAS Number: 16433-88-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 164.0 to 168.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...