Stampa…

1-Methylcyclohexanecarboxylic acid 99,0 GC, T

Fornitore: TCI
Attenzione

Synonyms: 1-Methyl-1-cyclohexanecarboxylic acid, 1-Methylcyclohexane-1-carboxylic acid

M0732-25G M0732-5G
TCIAM0732-25GEA 120 EUR
TCIAM0732-25G TCIAM0732-5G
1-Methylcyclohexanecarboxylic acid 99,0 GC, T
1-Methylcyclohexanecarboxylic acid
Formula: C₈H₁₄O₂
Peso molecolare: 142.2 g/mol
Punto di ebollizione: 236°C
Punto di fusione: 39
MDL Number: MFCD00001463
CAS Number: 1123-25-7
EINECS: 214-371-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d52e938324d9cf86f41e9a2f1e472a25351acf3d&vid=d52e938324d9cf86f41e9a2f1e472a25351acf3d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 36.0 to 40.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR