Stampa…
3,4-Dimethyl-1,2-cyclopentanedione ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms: 3,4-Dimethylcyclopentane-1,2-dione, 3,4-Dimethyl-1,2-cyclopentadione
D3782-1G D3782-25G
TCIAD3782-1GEA 16 EUR
TCIAD3782-1G TCIAD3782-25G
3,4-Dimethyl-1,2-cyclopentanedione ≥98.0% (by titrimetric analysis)
3,4-Dimethyl-1,2-cyclopentanedione
Formula: C₇H₁₀O₂
Peso molecolare: 126,16 g/mol
Punto di ebollizione: 219 °C (1013 hPa)
Punto di fusione: 71…72 °C
MDL Number: MFCD00143063
CAS Number: 13494-06-9
EINECS: 236-810-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6a9b2c681b95a5aaedadb9220b84c83760181e4&vid=f6a9b2c681b95a5aaedadb9220b84c83760181e4&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C