(Bromomethyl)cyclobutane ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Cyclobutyl methyl bromide
B3679-25G
B3679-5G
TCIAB3679-25GEA
167
EUR
TCIAB3679-25G
TCIAB3679-5G
(Bromomethyl)cyclobutane ≥98.0% (by GC)
(Bromomethyl)cyclobutane
Formula:
C₅H₉Br Peso molecolare: 149,03 g/mol Punto di ebollizione: 124°C Densità: 1,35 g/cm³ (20 °C) Flash Pt: 36°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00040960 CAS Number: 17247-58-4 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...