Stampa…

Trimethyloxonium tetrafluoroborate ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

T1507-25G T1507-5G
TCIAT1507-25GEA 106 EUR
TCIAT1507-25G TCIAT1507-5G
Trimethyloxonium tetrafluoroborate ≥95.0% (by titrimetric analysis)
Trimethyloxonium tetrafluoroborate
Formula: C₃H₉BF₄O
Peso molecolare: 147,91 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00011798
CAS Number: 420-37-1
EINECS: 206-994-4
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=094d8517ec898d8978dac4b96a9b6f517e913a7b&vid=094d8517ec898d8978dac4b96a9b6f517e913a7b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR