Stampa…

Chloro(dimethyl)(phenyl)silane ≥96.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Chlorodimethylphenylsilane

D1147-25ML D1147-5ML
TCIAD1147-25MLEA 62 EUR
TCIAD1147-25ML TCIAD1147-5ML
Chloro(dimethyl)(phenyl)silane ≥96.0% (by GC)
Chloro(dimethyl)(phenyl)silane
Formula: C₈H₁₁ClSi
Peso molecolare: 170,71 g/mol
Flash Pt: 82°C
MDL Number: MFCD00000499
CAS Number: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1268fe2b596dd5437377e191d895325c98c7b3c&vid=b1268fe2b596dd5437377e191d895325c98c7b3c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR