Stampa…

Dicyclohexyl phthala ≥99.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Dicyclohexyl phthalate

P0293-25G P0293-500G
TCIAP0293-25GEA 16 EUR
TCIAP0293-25G TCIAP0293-500G
Dicyclohexyl phthala ≥99.0% (by GC)
Dicyclohexyl phthala
Formula: C₂₀H₂₆O₄
Peso molecolare: 330,42 g/mol
MDL Number: MFCD00003849
CAS Number: 84-61-7
EINECS: 201-545-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7d152ef8d665b480e7ea91c9cdc6d5146b98e77&vid=d7d152ef8d665b480e7ea91c9cdc6d5146b98e77&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (GC) min. 99.0 %
Melting point 62.0 to 67.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR