Dibutyltin diacetate ≥95.0% (W)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Di-n-butyltin diacetate
Formula:
C₁₂H₂₄O₄Sn Peso molecolare: 351.03 g/mol Punto di ebollizione: 145°C Flash Pt: 143°C |
MDL Number:
MFCD00008697 CAS Number: 1067-33-0 EINECS: 213-928-8 UN: 2922 ADR: 8 II |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (Gravimetric Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...