Stampa…

1-Acetylpiperazine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A1399-25G A1399-5G
TCIAA1399-25GEA 81 EUR
TCIAA1399-25G TCIAA1399-5G
1-Acetylpiperazine ≥98.0% (by GC, titration analysis)
1-Acetylpiperazine
Formula: C₆H₁₂N₂O
Peso molecolare: 128,17 g/mol
MDL Number: MFCD00058676
CAS Number: 13889-98-0
EINECS: 237-659-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0bf8118b283906683c319c9803188f24ea89a74&vid=d0bf8118b283906683c319c9803188f24ea89a74&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR