4-Nitrophthalic anhydride ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₃NO₅ Punto di ebollizione: 197°C |
MDL Number:
MFCD00005922 CAS Number: 5466-84-2 EINECS: 226-776-2 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 97.0 % |
Melting point | 117.0 to 122.0 deg-C |
Solubility in Acetic acid | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...