Stampa…

3-Methylcrotonic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: 3,3-Dimethylacrylic acid, 3-Methylbut-2-enoic acid

M0543-25G M0543-500G M0543-100G
TCIAM0543-25GEA 16 EUR
TCIAM0543-25G TCIAM0543-500G TCIAM0543-100G
3-Methylcrotonic acid ≥98.0% (by GC, titration analysis)
3-Methylcrotonic acid
Formula: C₅H₈O₂
Peso molecolare: 100.12 g/mol
Punto di ebollizione: 98°C
MDL Number: MFCD00004366
CAS Number: 541-47-9
EINECS: 208-782-7
UN: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c748db36e1f74ef5742f60f68be5feba2ebf58e5&vid=c748db36e1f74ef5742f60f68be5feba2ebf58e5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 70.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR