Ethyl-3-methylcrotonate ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,3-Dimethylacrylic acid ethyl ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-methylbut-2-enoate, 3-Methyl-2-butenoic acid ethyl ester, 3-Methylcrotonic acid ethyl ester, Ethyl 3-Methyl-2-butenoate
M1441-25ML
M1441-5ML
TCIAM1441-25MLEA
73
EUR
TCIAM1441-25ML
TCIAM1441-5ML
Ethyl-3-methylcrotonate ≥97.0% (by GC)
Ethyl-3-methylcrotonate
Formula:
C₇H₁₂O₂ Peso molecolare: 128,17 g/mol Flash Pt: 43°C |
MDL Number:
MFCD00009187 CAS Number: 638-10-8 EINECS: 211-319-1 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...