Stampa…

Chloro(di-tert-butyl)phosphine ≥95.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Di-tert-butylchlorophosphine, Di-t-butylchlorophosphine

D3632-25G D3632-5G
TCIAD3632-25GEA 187 EUR
TCIAD3632-25G TCIAD3632-5G
Chloro(di-tert-butyl)phosphine ≥95.0% (by GC)
Chloro(di-tert-butyl)phosphine
Formula: C₈H₁₈ClP
Peso molecolare: 180,66 g/mol
Punto di ebollizione: 72°C
Punto di fusione: 3°C
Flash Pt: 62°C
MDL Number: MFCD00008815
CAS Number: 13716-10-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86697997b0448d9670fd1c5a26f0a5945a06f172&vid=86697997b0448d9670fd1c5a26f0a5945a06f172&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR