4-Picolyl bromide hydrobromide ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-(Bromomethyl)pyridine hydrobromide
B3315-5G
B3315-25G
TCIAB3315-5GEA
79
EUR
TCIAB3315-5G
TCIAB3315-25G
4-Picolyl bromide hydrobromide ≥97.0% (by titrimetric analysis)
4-Picolyl bromide hydrobromide
Formula:
C₆H₇Br₂N Peso molecolare: 252,94 g/mol |
MDL Number:
MFCD01863545 CAS Number: 73870-24-3 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 185.0 to 191.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...