1-Dodecylpyridinium chloride ≥98.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
N-Laurylpyridinium chloride, Dodecylpyridinium chloride
Formula:
C₁₇H₃₀ClN Peso molecolare: 283.88 g/mol |
MDL Number:
MFCD00011987 CAS Number: 104-74-5 EINECS: 203-232-2 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 93.0 % |
Water | max. 10.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...