Stampa…

DL-Tropic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid

T0533-25G T0533-250G
TCIAT0533-25GEA 31 EUR
TCIAT0533-25G TCIAT0533-250G
DL-Tropic acid ≥98.0% (by GC, titration analysis)
DL-Tropic acid
Formula: C₉H₁₀O₃
Peso molecolare: 166,18 g/mol
MDL Number: MFCD00004255
CAS Number: 552-63-6
EINECS: 209-020-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e46135a66941952f1a5128f424ac54e4d0c80d97&vid=e46135a66941952f1a5128f424ac54e4d0c80d97&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR