Stampa…

(±)-α-Terpineol ≥80.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

T0022-25ML T0022-500ML
TCIAT0022-25MLEA 18 EUR
TCIAT0022-25ML TCIAT0022-500ML
(±)-α-Terpineol ≥80.0% (by GC)
(±)-α-Terpineol
Formula: C₁₀H₁₈O
Peso molecolare: 154,25 g/mol
Flash Pt: 101°C
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6
Merck Index: 14,09171
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a2ae161197eb313a5f7642db02ac8ea0b2557bf&vid=8a2ae161197eb313a5f7642db02ac8ea0b2557bf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 80.0 %
Specific rotation [a]20/D -15.0 to -40.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR