Stampa…

Diethyl terephthalate ≥99.0% (by GC)

Fornitore: TCI

T0014-25G T0014-500G
TCIAT0014-25GEA 37 EUR
TCIAT0014-25G TCIAT0014-500G
Diethyl terephthalate ≥99.0% (by GC)
Diethyl terephthalate
Formula: C₁₂H₁₄O₄
Punto di ebollizione: 142°C
MDL Number: MFCD00039891
CAS Number: 636-09-9
EINECS: 211-249-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0d6c8134889d63af518e9364560504991edff7d&vid=f0d6c8134889d63af518e9364560504991edff7d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 43.0 to 47.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR