Ethyl-3-bromopropionate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Ethyl 3-bromopropanoate
B0647-25G
B0647-500G
B0647-100G
TCIAB0647-25GEA
43
EUR
TCIAB0647-25G
TCIAB0647-500G
TCIAB0647-100G
Ethyl-3-bromopropionate ≥98.0% (by GC)
Ethyl-3-bromopropionate
Formula:
C₅H₉BrO₂ Peso molecolare: 181,03 g/mol Flash Pt: 79°C |
MDL Number:
MFCD00000251 CAS Number: 539-74-2 EINECS: 208-724-0 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...