Cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
C0387-25ML
C0387-500ML
C0387-100ML
TCIAC0387-25MLEA
23
EUR
TCIAC0387-25ML
TCIAC0387-500ML
TCIAC0387-100ML
Cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Cyclopropanecarboxylic acid
Formula:
C₄H₆O₂ Peso molecolare: 86,09 g/mol Punto di ebollizione: 183°C Punto di fusione: 17…19 °C Densità: 1,087 g/cm³ (20 °C) Flash Pt: 71°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00001287 CAS Number: 1759-53-1 EINECS: 217-162-5 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...