2,4-Dimethylquinoline ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
D1237-1G
D1237-5G
TCIAD1237-1GEA
76
EUR
TCIAD1237-1G
TCIAD1237-5G
2,4-Dimethylquinoline ≥98.0% (by titrimetric analysis)
2,4-Dimethylquinoline
Formula:
C₁₁H₁₁N Peso molecolare: 157,22 g/mol Punto di ebollizione: 159°C |
MDL Number:
MFCD00006760 CAS Number: 1198-37-4 EINECS: 214-832-9 |
Risultati del test di specificazione
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...