Stampa…

7-Methylquinoline ≥98.0% (by GC)

Fornitore: TCI

M1180-25G M1180-5G
TCIAM1180-25GEA 0 EUR
TCIAM1180-25G TCIAM1180-5G
7-Methylquinoline ≥98.0% (by GC)
7-Methylquinoline
Formula: C₁₀H₉N
MDL Number: MFCD00006805
CAS Number: 612-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86a4fe44065c11ae65cb12b6254f037187f02a51&vid=d61b28322aa362ba30a1e31a73a49c3a05c8d5dc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 37.0 to 40.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR