Stampa…

6-Methoxyquinaldine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

M1414-25G M1414-5G
TCIAM1414-25GEA 429 EUR
TCIAM1414-25G TCIAM1414-5G
6-Methoxyquinaldine ≥98.0% (by GC, titration analysis)
6-Methoxyquinaldine
Formula: C₁₁H₁₁NO
Peso molecolare: 173,21 g/mol
Punto di ebollizione: 95 - 96 °C (0,5 -rr)
Punto di fusione: 67 °C
MDL Number: MFCD00006761
CAS Number: 1078-28-0
EINECS: 214-080-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a008840b6a1937cd8008b6189e10776fb4c4cff&vid=29c12e1bb9aa7144419b8e22603b384dc97488c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 64.0 to 68.0 deg-C
Solubility in Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR