7-Methyl-8-nitroquinoline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
M1864-5G
M1864-1G
TCIAM1864-5GEA
257
EUR
TCIAM1864-5G
TCIAM1864-1G
7-Methyl-8-nitroquinoline ≥98.0% (by GC, titration analysis)
7-Methyl-8-nitroquinoline
Formula:
C₁₀H₈N₂O₂ |
MDL Number:
MFCD00041234 CAS Number: 7471-63-8 EINECS: 231-268-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 183.0 to 187.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...